Order Entry
Italy
Orders LinkContactUsLinkComponent
PIG® CHAT MAT® Roll

PIG® CHAT MAT® Roll

Roll, heavyweight, 51,1 L, 1 roll/bag, 23 m×610 mm
Catalog # NEWPMAT619-DEC
Supplier:   NEW PIG
PIG® CHAT MAT® Roll
Catalog # NEWPMAT619-DEC
Supplier:  NEW PIG
PIG® CHAT MAT® Roll
PIG® CHAT MAT® Roll

Roll, heavyweight, 51,1 L, 1 roll/bag, 23 m×610 mm

Price
161,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Keep floors dry whilst delivering a message that can't be ignored.

  • Clearly visible message is repeated down the length of the roll to help create a safer environment for workers
  • Durable top layer prevents wear, even under heavy foot traffic
  • Under the Chat Mat top layer is the proven eight-layer, 100% polypropylene construction of PIG Mat
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Absorbs and retains oils, coolants, solvents and water
  • Flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job
  • Can be wrung out and incinerated after use to reduce waste or for fuels blending

Easy-tear perforations let you take only what you need, so you use less mat and save money. High-visibility yellow colour is easier to see in poorly lit areas to minimise tripping hazards; alerts workers to potential dangers and works in progress.

Specifications
  • Description:
    Roll, heavyweight
  • L×W:
    23 m×610 mm
  • Absorbency:
    51,1 L
  • Packed:
    1 roll/bag
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.