
ACY-738 95%
ACRO467911000
:
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
- Appearance:White to off-white Powder or solid
- HPLC:≥94%
- MS:complies
- NMR (1H-NMR):Consistent with structure
:
:270.29 g/mol
:GHS07
:Freezer
:1375465-91-0
:C₁₄H₁₄N₄O₂
:MFCD28347706