Order Entry
Italy
ContactUsLinkComponent
 
 
Anisole ≥99%, GPR RECTAPUR®
 
undefined
 
Anisole ≥99%, GPR RECTAPUR®
 

 

  • Assay :
    ≥ 99 %
  • Appearance :
    Clear colourless liquid
  • Odour:
    Characteristic
  • Density (20/4) :
    0,992 - 0,994

 

Frequently Bought Together