Order Entry
Italy
ContactUsLinkComponent
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride 94%, Sigma-Aldrich®
Irritant
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride 94%, Sigma-Aldrich®
Catalog # 407232-5G
CAS Number:  
undefined
Irritant
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline monohydrochloride 94%, Sigma-Aldrich®
Catalog # 407232-5G
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :359.29 g/mol
Melting Pt:224 °C (dec., lit.)
 :GHS07
Storage Temperature:Ambient
CAS Number:63857-00-1
Formula:C₂H₁₉ClN₂
MDL Number:MFCD00191759