Order Entry
Italy
ContactUsLinkComponent
2,6-Diphenyl-4H-thiopyran-4-one ≥96%, Sigma-Aldrich®
2,6-Diphenyl-4H-thiopyran-4-one ≥96%, Sigma-Aldrich®
Catalog # 454656-500MG
CAS Number:  
undefined
2,6-Diphenyl-4H-thiopyran-4-one ≥96%, Sigma-Aldrich®
Catalog # 454656-500MG
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :264.34 g/mol
Melting Pt:126…130 °C (lit.)
Storage Temperature:Ambient
CAS Number:1029-96-5
Formula:C₁₇H₁₂OS
MDL Number:MFCD00799527