Wright's stain solution, Sigma-Aldrich®
Catalog # 32857-1L
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Density:0.8 g/cm³
ADR:3,II
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
CAS Number:68988-92-1
:51938-72-8
MDL Number:MFCD00082143