Order Entry
Italy
ContactUsLinkComponent
2,4-Dichlorobenzenediazonium-1,5-naphthalenedisulphonate hydrate, Sigma-Aldrich®
Corrosive
2,4-Dichlorobenzenediazonium-1,5-naphthalenedisulphonate hydrate, Sigma-Aldrich®
Catalog # 320609-5G
CAS Number:  
undefined
Corrosive
2,4-Dichlorobenzenediazonium-1,5-naphthalenedisulphonate hydrate, Sigma-Aldrich®
Catalog # 320609-5G
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :461.29 g/mol (anhy)
Melting Pt:195 °C (dec., lit.)
ADR:8,II
 :GHS05
Storage Temperature:Refrigerator
CAS Number:123333-91-5
Formula:C₁₆H₁₀Cl₂N₂O₆S₂·xH₂O
MDL Number:MFCD08457580