Order Entry
Italy
ContactUsLinkComponent
Khellin for microscopy, Sigma-Aldrich®
Toxic
Khellin for microscopy, Sigma-Aldrich®
Catalog # 60730-10G
CAS Number:  
undefined
Toxic
Khellin for microscopy, Sigma-Aldrich®
Catalog # 60730-10G
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :260.25 g/mol
Melting Pt:150…154 °C (lit.)
ADR:6.1,III
 :GHS06
Storage Temperature:Ambient
CAS Number:82-02-0
Formula:C₁₄H₁₂O₅
MDL Number:MFCD00005007