Order Entry
Italy
ContactUsLinkComponent
Xylene fast yellow 2G 60% (dye content), Sigma-Aldrich®
Xylene fast yellow 2G 60% (dye content), Sigma-Aldrich®
Catalog # 201987-100G
CAS Number:  
undefined
Xylene fast yellow 2G 60% (dye content), Sigma-Aldrich®
Catalog # 201987-100G
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance (Color):
    Yellow
  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Wavelength:
    397- 403 nm
  • Extinction Coefficient:
    ≥14,000
  • Extinction Coefficient c = 0.02g/L; Water:
    ≥8,000
  • Carbon:
    ≥19.5 %
  • Nitrogen:
    ≥5.3 %

Specification Test Results

About this item

 :
 :551.28 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:228-819-0
CAS Number:6359-98-4
Formula:C₁₆H₁₂Cl₂N₄O₇S₂
MDL Number:MFCD00053367