Tetrachrome Stain (MacNeal), Sigma-Aldrich®
Catalog # 200190-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:81142-52-1
MDL Number:MFCD00134023