Order Entry
Italy
ContactUsLinkComponent
 
Direct Blue 71 50% (dye content), Sigma-Aldrich®
Catalog # 212407-50G
CAS Number:  
undefined
Direct Blue 71 50% (dye content), Sigma-Aldrich®
Catalog # 212407-50G
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :1029.87 g/mol
Storage Temperature:Ambient
CAS Number:4399-55-7
Formula:C₄H₂₃N₇O₁₃S₄
MDL Number:MFCD00003971