
Alizarin cyanin green F ≥60 % (dye content), Sigma-Aldrich®
Catalog # 214566-50G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:622.57 g/mol
Melting Pt:235…238 °C (lit.)
:GHS09
Storage Temperature:Ambient
CAS Number:4403-90-1
Formula:C₂₈H₂₂N₂O₈S₂
MDL Number:MFCD00001193