Order Entry
Italy
ContactUsLinkComponent
Indigotrisulphonic acid potassium salt ≥60% (by HPLC) ozone-scavenging reagent, Sigma-Aldrich®
Indigotrisulphonic acid potassium salt ≥60% (by HPLC) ozone-scavenging reagent, Sigma-Aldrich®
Certificates
undefined
Indigotrisulphonic acid potassium salt ≥60% (by HPLC) ozone-scavenging reagent, Sigma-Aldrich®

Specification Test Results

  • Appearance (Color) :
    Dark Blue to Very Dark Blue and Dark Purple to Very Dark Purple and Black
  • Appearance (Form) :
    Powder or Crystals or Powder with Chunk(s) or Crystals with Chunk(s)
  • Infrared spectrum :
    Conforms to Structure
  • Wavelength :
    597 - 603 nm
  • Extinction Coefficient :
    ≥ 15000
  • Extinction Coefficient ( c = 0.01g/l in H2O):
    ≥ 17000
  • Purity (HPLC):
    ≥ 60 %
  • Carbon Typically :
    19.5% - 26.0%
  • Nitrogen Typically :
    2.5% - 4.0%
  • Solubility (Color) c = 0.1%; Water:
    Dark Blue to Very Dark Blue

Specification Test Results

Frequently Bought Together