Order Entry
Italy
ContactUsLinkComponent
Gram color staining kit for tissue, Sigma-Aldrich®
Corrosive
Environment
Gram color staining kit for tissue, Sigma-Aldrich®
Catalog # HT90T-1KT
CAS Number:  
undefined
Corrosive
Environment
Gram color staining kit for tissue, Sigma-Aldrich®
Catalog # HT90T-1KT
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

About this item

 :
ADR:9,III
 :GHS05
 :GHS09
Storage Temperature:Ambient
 :1