0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Jenner's stain
Jenner's stain, Sigma-Aldrich®
Jenner's stain, Sigma-Aldrich®
Jenner's stain, Sigma-Aldrich®
Catalog # J1625-25G
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Expand image
Jenner's stain, Sigma-Aldrich®
Catalog # J1625-25G
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
Pk:
25 g
Seq:
3308
Specifications
About this item
Product Details
:
Storage Temperature:
Ambient
CAS Number:
62851-42-7
MDL Number:
MFCD00081733
Product Details