Jenner's stain, certified by the Biological Stain Commission, Sigma-Aldrich®
Catalog # J1875-25G
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:62851-42-7
MDL Number:MFCD00081733