Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Wright-Giemsa stain (solutions & kits)
Wright-Giemsa stain modified, Sigma-Aldrich®
Wright-Giemsa stain modified, Sigma-Aldrich®
Wright-Giemsa stain modified, Sigma-Aldrich®
Catalog # WG32-1L
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Expand image
Wright-Giemsa stain modified, Sigma-Aldrich®
Catalog # WG32-1L
Supplier:
SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Ambient
:
1
MDL Number:
MFCD00132593
Product Details
Documentation