Wright Stain Set Set for Hema-Tek® I, 1000 and 2000, Sigma-Aldrich®
Catalog # WSHT-1SET
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 SET
- Seq:0125
Specifications
About this item
:
ADR:3,II
:GHS02
:GHS06
:GHS08
Storage Temperature:Ambient
:1