N-(5-Aminopentyl)biotinamide trifluoroacetate salt, Sigma-Aldrich®
Catalog # A5348-25MG
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
- Environmentally Preferable:
Specifications
About this item
Synonyms: Biotin-cadaverine
:
:442.50 g/mol
Storage Temperature:Ambient
Formula:C₁₇H₂₉F₃N₄O₄S
MDL Number:MFCD03452814