Fontana-Masson Stain Kit, Sigma-Aldrich®
Catalog # HT200-1KT
Supplier: SIGMA ALDRICH MICROSCOPY
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
About this item
:
:M-Clarity™ quality level = MQ600 (more info.)
UN:3264
:GHS05
:GHS09
Storage Temperature:Refrigerator
:1