Zanamivir monohydrate
Glass bottle, 1 g
Codice catalogo ACRO462960010
Nome del fornitore: Thermo Fisher Scientific
Zanamivir monohydrate
Codice catalogo ACRO462960010
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 1260601-68-0
Jump to:
Product Details & Documents
- Bioactive
Synonyms: (2R,3R,4S)-3-Acetamido-4-guanidino-2-((1R,2R)-1,2,3-trihydroxypropyl)-3,4-dihydro-2H-pyran-6-carboxylic acid hydrate
- CAS Number:1260601-68-0
- Formula:C₁₂H₂₀N₄O₇·H₂O
- MDL Number:MFCD00866966
- Molecular Weight:350.33 g/mol
- Melting Pt:256 °C
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:4 % to 9 % (1 g, 105°C) (vacuum)
- HPLC:≥97.5 %
- Specific optical rotation:+36° to +38° (20°C, 589 nm) (1 % in water)
Specifications
- Pack type:Glass bottle
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



