Maleic anhydride, briquettes
: 2,5-Furanedione
:108-31-6
:
:C₄H₂O₃
:MFCD00005518
:200 °C (760 mmHg)
:52…55 °C
:1.48 g/cm³ (20 °C)
:98.06 g/mol
:102 °C
:Ambient
:8 III
:13,05727
:2215
:GHS05
:GHS07
:GHS08
- Environmentally Preferable:
- Appearance:White Briquettes
- Infrared spectrum:Conforms
- Melting point:50 to 56 °C
- Free acid:≤0.5 (as maleic acid)
- Titration with NaOH:98.5 to 102.5 %
Frequently Bought Together