

Dofetilide
:115256-11-6
:
:C₁₉H₂₇N₃O₅S₂
:MFCD00869707
:441.56 g/mol
:149…153 °C
:GHS07
:GHS08
:Ambient
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:150 to 157 °C
- Water:≤1 %
- Solubility:(2 % in DMSO) Clear solution
- HPLC:≥97.5 %
Frequently Bought Together