
5-Isopropyl-2-methylphenol
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:499-75-2
:
Formula:C₁₀H₁₄O
MDL Number:MFCD00002236
Boiling Pt:238 °C (760 mmHg)
Melting Pt:3 °C
Density:0.976
Flash Pt:106 °C
Storage Temperature:Ambient
UN:3145
ADR:8,III
:150.22 g/mol
:GHS07
Specification Test Results
- Appearance:Clear yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5220 to 1.5240 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together