Order Entry
Italy
Orders LinkContactUsLinkComponent
MCDh 4

MCDh 4

2,5 L
Catalog # RALA313600-2500
Supplier:   RAL
MCDh 4
Catalog # RALA313600-2500
Supplier:  RAL
CAS Number:  
MCDh 4
MCDh 4

2,5 L

CAS Number:  
Price
31,10 €
Log in for availability
1 * 2,5 L
Product Details & Documents

MCDh 4 - RINSING SOLUTION Water based - 6% ethanol

  • MCDh (Micro Chromatic Detection for Hematology)
  • for RAL Stainer Kit (Ref 360200)
  • Methanol-free

Role
- Removes excess stain from cells (better differentiation)
- Finalizes the staining

MCDh is a panoptic May-Grünwald Giemsa (MGG) type staining method. Methanol free, it is made of a sequence of four specific reagents use in blood and bone marrow cell differentiation and counting. This new generation of stains is in line with the standardisation trend in diagnostic techniques. Unlike more traditional stains, all MCDh reagents are fast and ready to use. As no dilution is needed, consistency and standardisation are guaranteed.

Components - Name (ref) Packing
MCDH 1 (Ref 313590) 1L - 2,5L
MCDH 2 (Ref 313570) 1L - 2,5L
MCDH 3 (Ref 313560) 1L - 2,5L
MCDH 4 (Ref 313600) 1L - 2,5L

Documents

Specifications

Specifications

  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.