Racks for cryoboxes, half box racks
Supplier: TENAK
Jump to:
About this item
Classic design of stainless steel rack for storage of cryoboxes in liquid nitrogen.
- Can be used with all brands of chest type freezers
- Suitable for all small cyroboxes with footprint 80×80 mm and height 50 mm
- The cryoboxes are secured with a locking rod
- Suitable for cryoboxes in cardboard, polypropylene and polycarbonate
Ordering information: Cryoboxes must be ordered separately.
7 Options Available Below
Product Details & Documents
Classic design of stainless steel rack for storage of cryoboxes in liquid nitrogen.
- Can be used with all brands of chest type freezers
- Suitable for all small cyroboxes with footprint 80×80 mm and height 50 mm
- The cryoboxes are secured with a locking rod
- Suitable for cryoboxes in cardboard, polypropylene and polycarbonate
Ordering information: Cryoboxes must be ordered separately.
Product Family Options
Product Information
- CompartmentsW×D×H ext.AvailabilityPrice