Order Entry
Italy
ContactUsLinkComponent
Heat resistant gloves, Duo-Therm A185
 :  Portwest
undefined
Heat resistant gloves, Duo-Therm A185
 :  Portwest
  • Catalog No:
  • PRTWA185O4RL
  • PRTWA185O4RXL
  • PRTWA185O4RXXL
  • PRTWA185Y4RXL
  • PRTWA185Y4RM
  • PRTWA185Y4RXXL
  • PRTWA185O4RM
  • PRTWA185Y4RL
  • Model:
  • A185
  • A185
  • A185
  • A185
  • A185
  • A185
  • A185
  • A185
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Liner material:
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Finish:
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Latex coating on palm and fingertips
  • Gauge:
  • 7
  • 7
  • 7
  • 7
  • 7
  • 7
  • 7
  • 7
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (1131X), EN 511 (X1X)
  • EN 388:2016 (1131X), EN 511 (X1X)
  • EN 388:2016 (1131X), EN 511 (X1X)
  • EN 388:2016 (1131X), EN 511 (X1X)
  • EN 388:2016 (1131X), EN 511 (X1X)
  • EN 388:2016 (1131X), EN 511 (X1X)
  • EN 388:2016 (1131X), EN 511 (X1X)
  • EN 388:2016 (1131X), EN 511 (X1X)
Frequently Bought Together