Analytical standard, Oxadixyl, reference material, Fluka™
Codice catalogo HONCR1743-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:77732-09-3
- Description:Oxadixyl
- Description:Oxadixyl
- Form:Neat