Analytical standard, Dimethenamid-P, reference material, Fluka™
HONCR1721-100MG
:
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:163515-14-8
- Description:Dimethenamid-P
- Storage temperature:2…8 °C
- Description:Dimethenamid-P
- Form:Neat