Order Entry
Italy
ContactUsLinkComponent
Analytical standard, Metolachlor, reference material, Fluka™
Analytical standard, Metolachlor, reference material, Fluka™
Catalog # HONCR1792-100MG
CAS Number:  
Analytical standard, Metolachlor, reference material, Fluka™
Catalog # HONCR1792-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Standards:
    Reference material
  • CAS No.:
    51218-45-2
  • Description:
    Metolachlor
  • Description:
    Metolachlor
  • Form:
    Neat

Specifications

About this item