Analytical standard, alpha-Endosulfan, reference material, Fluka™
Catalog # HONCR1772-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Standards:Reference material
- CAS No.:959-98-8
- Description:alpha-Endosulfan
- Formula:C₉H₆Cl₆O₃S
- Storage temperature:Ambient
- Description:alpha-Endosulfan
- Form:Neat