Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Aspergillus
oryzae
Protease
Aspergillus oryzae Protease
Aspergillus oryzae Protease
Catalog # TCIAP0538-25G
Supplier:
TCI
CAS Number:
Expand image
Aspergillus oryzae Protease
Catalog # TCIAP0538-25G
Supplier:
TCI
CAS Number:
Specifications
Pk:
25 g
Species:
Aspergillus oryzae
Enzyme name:
Protease
Specifications
About this item
Product Details
Documentation
Product Details
Documentation