Order Entry
Italy
ContactUsLinkComponent
Pig Pepsin (from Stomach)
Pig Pepsin (from Stomach)
Catalog # TCIAP0103-25G
Supplier:  TCI
CAS Number:  
Pig Pepsin (from Stomach)
Catalog # TCIAP0103-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Source:
    Stomach
  • Species:
    Pig
  • Enzyme name:
    Pepsin

Specifications

About this item