0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Enzymes
Aspergillus
oryzae
Diastase
Aspergillus oryzae Diastase
TCIAD1115-25G
:
TCI
:
Aspergillus oryzae Diastase
TCIAD1115-25G
:
TCI
:
Pk:
25 g
Species:
Aspergillus oryzae
Enzyme name:
Diastase