0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Enzymes
Bovine catalase (from Liver)
Bovine catalase (from Liver)
Catalog # TCIAC0052-1G
Supplier:
TCI
CAS Number:
Expand image
Bovine catalase (from Liver)
Catalog # TCIAC0052-1G
Supplier:
TCI
CAS Number:
Specifications
Pk:
1 g
Source:
Liver
Species:
Bovine
Enzyme name:
Catalase
Specifications
About this item
Product Details
Product Details