0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
ContactUsLinkComponent
Enzymes
Yeast alcohol dehydrogenase
Yeast alcohol dehydrogenase
TCIAA0200-10MG
:
TCI
:
Yeast alcohol dehydrogenase
TCIAA0200-10MG
:
TCI
:
Pk:
10 mg
CAS No.:
9031-72-5
Species:
Yeast
Storage conditions:
Freezer
Form:
Crystals
Enzyme name:
Alcohol dehydrogenase
for Blood alcohol-test