

2,4,6-Trichloro-1,3,5-triazine for synthesis, Sigma-Aldrich®
Catalog # 8.02815.5000
Supplier: Merck
CAS Number:
Specifications
- Pk:5 kg
- Seq:4114
Specifications
Specification Test Results
- Solubility:(20 °C) (decomposition)
- Melting Point:144 - 146 °C
- Molar Mass:184.41 g/mol
- Bulk Density:600 - 900 kg/m3
- Boiling Point:193 - 198 °C (1013 hPa)
- Vapor Pressure:0.01 hPa (20 °C)
- Flash Point:190 °C
- Density:1.92 g/cm3 (20 °C)
- pH:(H2O) acidic,Hydrolysis
- Ignition Point:650 °C
Specification Test Results
About this item
:
:184.41 g/mol
Boiling Pt:190 °C (1013 hPa)
Density:1.32 g/cm³ (20 °C)
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:203-614-9
UN:2670
:GHS06
:GHS05
CAS Number:108-77-0
Formula:C₃Cl₃N₃
MDL Number:MFCD00006046