


Zinc phosphide ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1314-84-7
:
Formula:P₂Zn₃
MDL Number:MFCD00049631
:258.12 g/mol
Merck Index:14,10152
Boiling Pt:1100 °C
Melting Pt:420 °C
Density:4.55
Storage Temperature:Ambient
EINECS:215-244-5
UN:1714
ADR:4.3,I
:GHS02
:GHS06
:GHS09
Related Information
Frequently Bought Together