Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Hydroxy-1-naphthaldehyde

2-Hydroxy-1-naphthaldehyde

Glass bottle for solids, 25 g
Catalog # ICNA0215129825
Supplier:   MP Biomedicals
2-Hydroxy-1-naphthaldehyde
Catalog # ICNA0215129825
Supplier:  MP Biomedicals
CAS Number:  708-06-5
2-Hydroxy-1-naphthaldehyde

Glass bottle for solids, 25 g

CAS Number:  708-06-5
Price
23,10 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Hydroxy-α-naphthaldehyde
  • CAS Number:708-06-5
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00004005
  • Molecular Weight:172.18 g/mol
  • EINECS:211-902-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.