
1-Bromo-2-naphthol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:573-97-7
:
Formula:C₁₀H₇BrO
MDL Number:MFCD00003869
:223.07 g/mol
Boiling Pt:314 °C (1013 hPa)
Melting Pt:76…81 °C
Storage Temperature:Ambient
EINECS:209-363-1
:GHS07
Related Information
Frequently Bought Together