
Tetradecyltrimethylammonium bromide 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1119-97-7
:8044-71-1
Formula:C₁₇H₃₈BrN
MDL Number:MFCD00011770
:336.394 g/mol
Merck Index:12,06419
Melting Pt:245…250 °C
Storage Temperature:Ambient
EINECS:214-291-9
UN:1759
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:245 to 250 °C
- Titration Argentometric:98.5 to 101.5 %
Specification Test Results
Frequently Bought Together