

Dodecyltrimethylammonium bromide for synthesis
: Merck
:1119-94-4
:
:C₁₅H₃₄BrN
:MFCD00011767
:308.35 g/mol
:264…266 °C
:Ambient
:214-290-3
:2811
:6.1,II
:M-Clarity™ quality level = MQ200
:GHS06
:GHS09
- Melting Point:237 - 245 °C
- Molar Mass:308.34 g/mol
- Hill Formula:C15H34BrN
Frequently Bought Together