
Gallium trifluoride, anhydrous ≥99.85% (metals basis)
Catalog # 32112.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:126.72 g/mol
Merck Index:14,4354
Boiling Pt:∼1000 °C
Melting Pt:800 °C (subl.)
Density:4.47
Storage Temperature:Ambient
EINECS:232-004-5
UN:3260
ADR:8,III
:GHS05
CAS Number:7783-51-9
Formula:F₃Ga
MDL Number:MFCD00011021