Sodium hexametaphosphate 66.5% P2O5 min., tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium metaphosphate, SHMP
CAS Number:10124-56-8
Formula:Na₆P₆O₁₈
MDL Number:MFCD00136045
:611.77 g/mol
Boiling Pt:1500 °C
Melting Pt:628 °C
Density:1.25 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-343-1
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together