Order Entry
Italy
ContactUsLinkComponent
Potassium chloride 99.0-100.5% USP, FCC, Macron Fine Chemicals™
Potassium chloride 99.0-100.5% USP, FCC, Macron Fine Chemicals™
Catalog # 6838-20
CAS Number:  
undefined
Potassium chloride 99.0-100.5% USP, FCC, Macron Fine Chemicals™
Catalog # 6838-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets U.S.P Requirements:
  • Meets F.C.C. Requirements:
  • GMP Manufactured Product:
  • Food GMP Manufactured Product:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Food Chemical:
  • Bulk Pharmaceutical Chemical:
  • USP - Assay (KCl) (by Ag titrn):
    99.0 - 100.5 %
  • USP - Acidity or Alkalinity (no pink color then pink):
    Passes Test
  • USP - Bromide (Br)(USP):
    ≤0.1 %
  • USP - Calcium and Magnesium:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Iodide (I)(USP):
    ≤0.005 %
  • USP - Loss on Drying at 105°C:
    ≤1.0 %
  • USP - Sodium (Na):
    Passes Test
  • FCC - Assay (KCl) (by Ag titrn):
    99.0 - 100.5 %
  • FCC - Acidity or Alkalinity (no pink color then pink):
    Passes Test
  • FCC - Bromide and/or Iodide (FCC):
    Passes Test
  • FCC - Heavy Metals (as Pb)(FCC):
    ≤5 mg/kg
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Loss on Drying at 105°C:
    ≤1.0 %
  • FCC - Sodium (Na):
    Passes Test
  • Solubility (10 in 50):
    Passes Test

Specification Test Results

About this item

 :
 :74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360