
meta-Phosphoric acid ~100%, lumps (glassy), Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:37267-86-0
:
Formula:(HPO₃)n
MDL Number:MFCD00011351
:79.98 g/mol
Boiling Pt:600 °C (1.013 hPa)
Melting Pt:200 °C
Density:2.0 g/cm³
Storage Temperature:Ambient
EINECS:253-433-4
UN:3260
ADR:8,II
:GHS05
Specification Test Results
- Assay of (HPO₃)n:40 - 50 %
- Assay of (naPO₃)n:50 - 60 %
- Arsenic (As):Max. 0.0005 %
- Iron (Fe):Max. 0.01 %
- Heavy metals (as Pb):Max. 0.001 %
- Chloride (Cl):Max. 0.002 %
Specification Test Results
Frequently Bought Together