Order Entry
Italy
ContactUsLinkComponent
meta-Phosphoric acid, GPR RECTAPUR®
Corrosive
meta-Phosphoric acid, GPR RECTAPUR®
Catalog # 291903W
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
meta-Phosphoric acid, GPR RECTAPUR®
Catalog # 291903W
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Identification:
    Passes test
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 20 ppm
  • Pb (Lead):
    Max. 20 ppm

Specification Test Results

About this item

 :
 :79.98 g/mol
Boiling Pt:260 °C (1013 hPa)
Melting Pt:21 °C
Density:2.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:253-433-4
UN:3260
ADR:8,III
 :GHS05
CAS Number:37267-86-0
Formula:(HPO₃)n
MDL Number:MFCD00011351