Order Entry
Italy
ContactUsLinkComponent
Phenazone
Irritant
Phenazone
Catalog # EHERC16003100
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Phenazone
Catalog # EHERC16003100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :188.23 g/mol
Merck Index:12,00757
Boiling Pt:388 °C (1013 hPa)
Melting Pt:152…154 °C
Density:1.25 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-486-6
 :GHS07
CAS Number:60-80-0
Formula:C₁₁H₁₂N₂O
MDL Number:MFCD00003146