Order Entry
Italy
ContactUsLinkComponent
2-tert-Butyl-p-cresol 99%
Corrosive
Environment
2-tert-Butyl-p-cresol 99%
Certificates
undefined
Corrosive
Environment
2-tert-Butyl-p-cresol 99%

Specification Test Results

  • Appearance:
    White to yellow to light beige Crystalline mass
  • Infrared spectrum:
    Conforms
  • Melting point:
    50 to 53 °C
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together