Order Entry
Italy
Orders LinkContactUsLinkComponent
4-Heptylphenol

4-Heptylphenol

Glass bottle for solids, 1 g
Catalog # MAYBJFD01551.1
4-Heptylphenol
Catalog # MAYBJFD01551.1
Supplier:  
CAS Number:  1987-50-4
4-Heptylphenol

Glass bottle for solids, 1 g

CAS Number:  1987-50-4
Discontinued
Product Details & Documents
Synonyms:  4-n-Heptylphenol
  • CAS Number:1987-50-4
  • Formula:C₁₃H₂₀O
  • MDL Number:MFCD00041751
  • Molecular Weight:192.3 g/mol
  • Boiling Pt:298 °C (1013 hPa)
  • Melting Pt:26…28 °C
  • Density:0.939 g/cm³ (20 °C)
  • EINECS:217-862-0
  • UN:2430
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.